missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxydodecanoic Acid Methyl Ester, TRC

Product Code. 30390393
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390393 100 mg
30402875 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30390393 Supplier Toronto Research Chemicals Supplier No. TRCH939600100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72864-23-4
Molecular Formula C13 H26 O3
Molecular Weight (g/mol) 230.34
Synonym (±)-Methyl 3-Hydroxydodecanoate, Methyl 3-Hydroxydodecanoate, Methyl β-Hydroxydodecanoate
IUPAC Name methyl 3-hydroxydodecanoate
SMILES CCCCCCCCCC(O)CC(=O)OC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H26O3/c1-3-4-5-6-7-8-9-10-12(14)11-13(15)16-2/h12,14H,3-11H2,1-2H3
Formula Weight 230.1882
Chemical Name or Material 3-Hydroxydodecanoic Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.