missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxydodecanoic acid, MedChemExpress

Product Code. 16427477
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16427477 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16427477 Supplier Medchem Express Supplier No. HY113107

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 3-Hydroxydodecanoic acid is a medium-chain fatty acid associated with fatty acid metabolic disorders.

3-Hydroxydodecanoic acid is a medium-chain fatty acid associated with fatty acid metabolic disorders.

Specifications

Chemical Name or Material 3-Hydroxydodecanoic acid
Physical Form Solid
CAS 1883-13-2
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C12H24O3
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information DMSO : 100 mg/mL (462.28 mM; Need ultrasonic)
SMILES CCCCCCCCCC(O)CC(O)=O
Molecular Weight (g/mol) 216.32
Formula Weight 216.32
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.