missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxychlorpromazine Hydrochloride, TRC

Product Code. 30400941
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400941 1 mg -
30403814 5 mg -
30391467 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30400941 Supplier Toronto Research Chemicals Supplier No. TRCH9506201MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17800-28-1
Molecular Formula C17 H19 Cl N2 O S . H Cl
Molecular Weight (g/mol) 371.325
Synonym 2-Chloro-10-[3-(dimethylamino)propyl]-10H-phenothiazin-3-ol Hydrochloride, 2-Chloro-10-[3-(dimethylamino)propyl]-phenothiazin-3-ol Hydrochloride
IUPAC Name 2-chloro-10-[3-(dimethylamino)propyl]phenothiazin-3-ol;hydrochloride
SMILES Cl.CN(C)CCCN1c2ccccc2Sc3cc(O)c(Cl)cc13

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C17H19ClN2OS.ClH/c1-19(2)8-5-9-20-13-6-3-4-7-16(13)22-17-11-15(21)12(18)10-14(17)20;/h3-4,6-7,10-11,21H,5,8-9H2,1-2H3;1H
Formula Weight 370.067
Chemical Name or Material 3-Hydroxychlorpromazine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.