Learn More
3-Hydroxybutyric acid, tech.
Descripción
Industrially, 3-hydroxybutanoic acid is used for the manufacture of biodegradable plastics. It is also acts as a solvent in organoleptics, cosmetics. It is also used in enantioselective synthesis of D-(-)-3-hydroxybutyric acid by Escherichia coli.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
| Color | Colorless |
| Density | 1.13 g/mL |
| Boiling Point | 130°C (12 mmHg) |
| Flash Point | >112°C (234°F) |
| Refractive Index | 1.4445 |
| Quantity | 5 g |
| Sensitivity | Hygroscopic |
| Merck Index | 14,4816 |
| Solubility Information | Soluble in water, alcohol, ether |
| Formula Weight | 104.1 |
| Mostrar más |
Seguridad y manipulación
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.