missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybutyric acid, MedChemExpress

Product Code. 16495576
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16495576 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16495576 Supplier Medchem Express Supplier No. HY113378
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 3-Hydroxybutyric acid (β-Hydroxybutyric acid) is a metabolite that is elevated in type I diabetes. 3-Hydroxybutyric acid can modulate the properties of membrane lipids.

3-Hydroxybutyric acid (β-Hydroxybutyric acid) is a metabolite that is elevated in type I diabetes. 3-Hydroxybutyric acid can modulate the properties of membrane lipids.

Specifications

Chemical Name or Material 3-Hydroxybutyric acid
Physical Form Solid
CAS 300-85-6
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C4H8O3
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym β-Hydroxybutyric acid
Solubility Information H2O : 250 mg/mL (2401.54 mM; Need ultrasonic) ∣0.1 M NaOH : 100 mg/mL (960.61 mM; ultrasonic and adjust pH to 6 with NaOH)
SMILES CC(O)CC(O)=O
Molecular Weight (g/mol) 104.1
Formula Weight 104.1
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.