missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzylhydrazine dihydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 10378360 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10378360 5 g Glass bottle 5g
10687392 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10378360 Supplier Thermo Scientific Acros Supplier No. 229030050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81012-99-9
Molecular Formula C7H12Cl2N2O
Molecular Weight (g/mol) 211.09
MDL Number MFCD00044695
InChI Key ONOJPUDFIOEGCX-UHFFFAOYSA-N
Synonym 3-hydroxybenzylhydrazine dihydrochloride, 3-hydrazinylmethyl phenol dihydrochloride, alpha-hydrazino-m-cresol dihydrochloride, 3-hydroxybenzylhydrazine 2hcl, 3-hydrazinomethyl phenol dihydrochloride, 3-hydroxybenzylhydrazine hydrochloride, m-hydroxybenzylhydrazine dihydrochloride, 3-hydrazinomethyl-phenol dihydrochloride, phenol, 3-hydrazinomethyl-, dihydrochloride, 3-hydrazinomethyl phenol, chloride, chloride
PubChem CID 2724402
IUPAC Name 3-(hydrazinylmethyl)phenol;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].NNCC1=CC=CC(O)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 138°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HOC6H4CH2NHNH2·2HCl
Quantity 5 g
Formula Weight 211.09
Percent Purity 98%
Physical Form Adhering Powder
Chemical Name or Material 3-Hydroxybenzylhydrazine dihydrochloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Hydroxybenzylhydrazine dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.