missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzyl Alcohol, 97%

Product Code. p-3736590 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10275300 5 g Glass bottle 5g
10091100 25 g Glass bottle 25g
10265350 100 g Glass bottle 100g
10608693 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10275300 Supplier Thermo Scientific Acros Supplier No. 121020050
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 620-24-6 | C7H8O2 | 124.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-24-6
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00004643
InChI Key OKVJCVWFVRATSG-UHFFFAOYSA-N
Synonym 3-hydroxybenzyl alcohol, 3-hydroxymethyl phenol, benzenemethanol, 3-hydroxy, 3-methylolphenol, m-hydroxybenzyl alcohol, 3-hydroxybenzenemethanol, benzyl alcohol, m-hydroxy, 3-hydroxybenzylalcohol, 3-oh-benzyl-alcohol, 3-hydroxymethyl-phenol
PubChem CID 102
ChEBI CHEBI:17069
IUPAC Name 3-(hydroxymethyl)phenol
SMILES C1=CC(=CC(=C1)O)CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 67°C to 71°C
Color Beige to Brown or Pink
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula HOC6H4CH2OH
Quantity 5 g
Beilstein 06, 896
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in chloroform,soluble in ethanol and ether
Formula Weight 124.14
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxybenzyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Hydroxybenzyl alcohol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.