missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzaldehyde, TRC

Product Code. 17992495
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
Unit Size:
100g
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17992495 5 g 5g
17924845 250 g 250g
17921915 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17992495 Supplier Toronto Research Chemicals Supplier No. TRCH8289655G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-83-4
Molecular Formula C7 H6 O2
Molecular Weight (g/mol) 122.12
Synonym Benzaldehyde, 3-hydroxy-, Benzaldehyde, m-hydroxy- (8CI), 3-Hydroxybenzaldehyde, 3-Formylphenol, 5-Hydroxybenzaldehyde, NSC 3504, m-Formylphenol, m-Hydroxybenzaldehyde
IUPAC Name 3-hydroxybenzaldehyde
SMILES Oc1cccc(C=O)c1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H6O2/c8-5-6-2-1-3-7(9)4-6/h1-5,9H
Formula Weight 122.0368
Chemical Name or Material 3-Hydroxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.