missing translation for 'onlineSavingsMsg'
Learn More

3'-Hydroxyacetophenone, 99+%, Thermo Scientific Chemicals

Product Code. 10687201
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-71-1
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002298
InChI Key LUJMEECXHPYQOF-UHFFFAOYSA-N
Synonym 3'-hydroxyacetophenone, 3-hydroxyacetophenone, 1-3-hydroxyphenyl ethanone, m-hydroxyacetophenone, 3-acetylphenol, 1-3-hydroxyphenyl ethan-1-one, ethanone, 1-3-hydroxyphenyl, m-acetylphenol, 3-hydroxy acetophenone, 3-acetophenol
PubChem CID 8487
IUPAC Name 1-(3-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 94.0°C to 97.0°C
Color Beige to Brown
Boiling Point 296.0°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 08, 86
Formula Weight 136.15
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 3'-Hydroxyacetophenone, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'