missing translation for 'onlineSavingsMsg'
Learn More

3'-Hydroxy Stanozolol, TRC

Product Code. 30393222
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393222 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393222 Supplier Toronto Research Chemicals Supplier No. TRCH9535305MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3'-Hydroxystanozolol
CAS 2295-92-3
Molecular Formula C21 H32 N2 O2
Quantity 5 mg
Synonym (5alpha,17beta)-17-Methyl-1'H-Androstano[3,2-c]pyrazole-5',17-diol, 1,2,3,3a,3b,4,5,5a,6,8,10,10a,10b,11,12,12a-Hexadecahydro-1,10a,12a-trimethylcyclopenta[7,8]phenanthro[2,3-c]pyrazole-1,9-diol, 1'H-Androstano[3,2-c]pyrazole-5',17-diol Cyclopenta[7,8]phenanthro[2,3-c]pyrazole deriv.
InChI Formula InChI=1S/C21H32N2O2/c1-19-11-14-17(22-23-18(14)24)10-12(19)4-5-13-15(19)6-8-20(2)16(13)7-9-21(20,3)25/h12-13,15-16,25H,4-11H2,1-3H3,(H2,22,23,24)/t12-,13+,15-,16-,19-,20-,21-/m0/s1
SMILES C[C@]1(O)CC[C@H]2[C@@H]3CC[C@H]4Cc5[nH]nc(O)c5C[C@]4(C)[C@H]3CC[C@]12C
Molecular Weight (g/mol) 344.49
Formula Weight 344.2464
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.