missing translation for 'onlineSavingsMsg'
Learn More

3’-Hydroxy Stanozolol Glucuronide Sodium Salt (Technical Grade), TRC

Product Code. 30399254
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399254 10 mg 10mg
30406258 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30399254 Supplier Toronto Research Chemicals Supplier No. TRCH95867010MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3’-Hydroxy Stanozolol Glucuronide Sodium Salt (Technical Grade)
Analyte or Component Names 3’-Hydroxy Stanozolol Glucuronide Sodium Salt (Technical Grade)
Shipping Condition Room Temperature
Molecular Formula C27 H39 N2 Na O8
Synonym (5alpha,17beta)-17-Hydroxy-17-methyl-2'H-androst-2-eno[3,2-c]pyrazol-5'-yl beta-D-Glucopyranosiduronic Acid Sodium Salt
InChI Formula InChI=1S/C27H40N2O8.Na/c1-25-11-14-17(28-29-22(14)37-24-20(32)18(30)19(31)21(36-24)23(33)34)10-12(25)4-5-13-15(25)6-8-26(2)16(13)7-9-27(26,3)35;/h12-13,15-16,18-21,24,30-32,35H,4-11H2,1-3H3,(H,28,29)(H,33,34);/q;+1/p-1/t12-,13+,15-,16-,18-,19-,20+,21-,24-,25-,26-,27-;/m0./s1
Solution Type Neat
SMILES C[C@]1(O)CC[C@H]2[C@@H]3CC[C@H]4Cc5[nH]nc(O[C@@H]6O[C@@H]([C@@H](O)[C@H](O)[C@H]6O)C(=O)O[Na])c5C[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name [(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[[(1S,2S,10S,13R,14S,17S,18S)-17-hydroxy-2,17,18-trimethyl-6,7-diazapentacyclo[11.7.0.02,10.04,8.014,18]icosa-4(8),5-dien-5-yl]oxy]oxane-2-carbonyl]oxysodium
Industry Type Forensics
Molecular Weight (g/mol) 542.597
Quantity 10 mg
Formula Weight 542.26
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.