missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3’-Hydroxy Stanozolol Glucuronide Sodium Salt (Technical Grade), TRC
Specifications
Specifications
| Chemical Name or Material | 3’-Hydroxy Stanozolol Glucuronide Sodium Salt (Technical Grade) |
| Analyte or Component Names | 3’-Hydroxy Stanozolol Glucuronide Sodium Salt (Technical Grade) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C27 H39 N2 Na O8 |
| Synonym | (5alpha,17beta)-17-Hydroxy-17-methyl-2'H-androst-2-eno[3,2-c]pyrazol-5'-yl beta-D-Glucopyranosiduronic Acid Sodium Salt |
| InChI Formula | InChI=1S/C27H40N2O8.Na/c1-25-11-14-17(28-29-22(14)37-24-20(32)18(30)19(31)21(36-24)23(33)34)10-12(25)4-5-13-15(25)6-8-26(2)16(13)7-9-27(26,3)35;/h12-13,15-16,18-21,24,30-32,35H,4-11H2,1-3H3,(H,28,29)(H,33,34);/q;+1/p-1/t12-,13+,15-,16-,18-,19-,20+,21-,24-,25-,26-,27-;/m0./s1 |
| Solution Type | Neat |
| SMILES | C[C@]1(O)CC[C@H]2[C@@H]3CC[C@H]4Cc5[nH]nc(O[C@@H]6O[C@@H]([C@@H](O)[C@H](O)[C@H]6O)C(=O)O[Na])c5C[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC Name | [(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[[(1S,2S,10S,13R,14S,17S,18S)-17-hydroxy-2,17,18-trimethyl-6,7-diazapentacyclo[11.7.0.02,10.04,8.014,18]icosa-4(8),5-dien-5-yl]oxy]oxane-2-carbonyl]oxysodium |
| Industry Type | Forensics |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?