missing translation for 'onlineSavingsMsg'
Learn More

3' '-Hydroxy Simvastatin Acid Sodium Salt, TRC

Product Code. 30392760
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392760 2.5 mg 2.5mg
30404014 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30392760 Supplier Toronto Research Chemicals Supplier No. TRCH9535152.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1316302-95-0
Molecular Formula C25H39NaO7
Molecular Weight (g/mol) 474.56
Synonym 1-Naphthaleneheptanoic acid, 1,2,6,7,8,8a-hexahydro-β,δ-dihydroxy-8-(3-hydroxy-2,2-dimethyl-1-oxobutoxy)-2,6-dimethyl-, sodium salt (1:1), (βR,δR,1S,2S,6R,8S,8aR)-, (βR,δR,1S,2S,6R,8S,8aR)-1,2,6,7,8,8a-Hexahydro-β,δ-dihydroxy-8-(3-hydroxy-2,2-dimethyl-1-oxobutoxy)-2,6-dimethyl-1-naphthaleneheptanoic acid sodium salt, 3''-Hydroxy simvastatin acid sodium salt, 3''-Hydroxysimvastatin hydroxy acid sodium salt, Sodium 3''-Hydroxytenivastatin, (3R,5R)-7-[(1S,2S,6R,8S,8aR)-8-[(2,2-Dimethyl-3-hydroxybutanoyl)oxy]-2,6-dimethyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoic acid monosodium salt
SMILES O=C(C(C)(C)C(O)C)O[C@H]1C[C@@H](C)C=C2[C@@]1([H])[C@](CC[C@@](C[C@@H](O)CC(O[Na])=O)([H])O)([H])[C@@H](C)C=C2

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C25H40O7.Na/c1-14-10-17-7-6-15(2)20(9-8-18(27)12-19(28)13-22(29)30)23(17)21(11-14)32-24(31)25(4,5)16(3)26;/h6-7,10,14-16,18-21,23,26-28H,8-9,11-13H2,1-5H3,(H,29,30);/q;+1/p-1/t14-,15-,16?,18+,19+,20-,21-,23-;/m0./s1
Formula Weight 474.2593
Chemical Name or Material 3''-Hydroxy Simvastatin Acid Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.