missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3’-Hydroxy Repaglinide (Mixture of Diastereomers), TRC
Specifications
Specifications
| Chemical Name or Material | 3'-hydroxy Repaglinide (mixture of diastereomers) |
| CAS | 874908-14-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C27 H36 N2 O5 |
| Quantity | 1 mg |
| Synonym | Benzoic acid, 2-ethoxy-4-[2-[[1-[2-(3-hydroxy-1-piperidinyl)phenyl]-3-methylbutyl]amino]-2-oxoethyl]- (9CI, ACI), 2-Ethoxy-4-[2-[[1-[2-(3-hydroxy-1-piperidinyl)phenyl]-3-methylbutyl]amino]-2-oxoethyl]benzoic acid (ACI) |
| InChI Formula | InChI=1S/C27H36N2O5/c1-4-34-25-15-19(11-12-22(25)27(32)33)16-26(31)28-23(14-18(2)3)21-9-5-6-10-24(21)29-13-7-8-20(30)17-29/h5-6,9-12,15,18,20,23,30H,4,7-8,13-14,16-17H2,1-3H3,(H,28,31)(H,32,33) |
| SMILES | CCOc1cc(CC(=O)NC(CC(C)C)c2ccccc2N3CCCC(O)C3)ccc1C(=O)O |
| IUPAC Name | 2-ethoxy-4-[2-[[1-[2-(3-hydroxypiperidin-1-yl)phenyl]-3-methylbutyl]amino]-2-oxoethyl]benzoic acid |
| Molecular Weight (g/mol) | 468.59 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?