missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-O-methyl-L-tyrosine Hydrochloride, TRC

Product Code. 30402005
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402005 50 mg -
30407956 100 mg -
30404921 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402005 Supplier Toronto Research Chemicals Supplier No. TRCH80505350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37466-29-8
Molecular Formula C10 H13 N O4 . H Cl
Molecular Weight (g/mol) 247.675
Synonym 4-O-Methyldopa Hydrochloride
IUPAC Name (2S)-2-amino-3-(3-hydroxy-4-methoxy-phenyl)propanoic acid;hydrochloride
SMILES Cl.COc1ccc(C[C@H](N)C(=O)O)cc1O

Specifications

InChI Formula InChI=1S/C10H13NO4.ClH/c1-15-9-3-2-6(5-8(9)12)4-7(11)10(13)14;/h2-3,5,7,12H,4,11H2,1H3,(H,13,14);1H/t7-;/m0./s1
Quantity 50 mg
Formula Weight 247.0611
Shipping Condition Room Temperature
Chemical Name or Material 3-Hydroxy-O-methyl-L-tyrosine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.