missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxy-N-aminoethyl-N’-aminocaproyldihydrocinnamoyl Cyclopamine (>90%), TRC
Specifications
Specifications
| Chemical Name or Material | 3-Hydroxy-N-aminoethyl-N’-aminocaproyldihydrocinnamoyl Cyclopamine |
| Analyte or Component Names | 3-Hydroxy-N-aminoethyl-N’-aminocaproyldihydrocinnamoyl Cyclopamine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C44 H65 N3 O4 |
| InChI Formula | InChI=1S/C44H65N3O4/c1-29-25-39-42(47(28-29)24-23-46-40(49)13-9-6-10-22-45-41(50)17-14-32-11-7-5-8-12-32)31(3)44(51-39)21-19-35-36-16-15-33-26-34(48)18-20-43(33,4)38(36)27-37(35)30(44)2/h5,7-8,11-12,15,29,31,34-36,38-39,42,48H,6,9-10,13-14,16-28H2,1-4H3,(H,45,50)(H,46,49)/t29-,31+,34-,35?,36?,38?,39+,42-,43-,44?/m0/s1 |
| Solution Type | Neat |
| SMILES | C[C@H]1C[C@H]2OC3(CCC4C5CC=C6C[C@@H](O)CC[C@]6(C)C5CC4=C3C)[C@H](C)[C@@H]2N(CCNC(=O)CCCCCNC(=O)CCc7ccccc7)C1 |
| IUPAC Name | N-[2-[(3S,3'R,3'aS,6'S,7'aR,11bR)-3-hydroxy-3',6',10,11b-tetramethylspiro[2,3,4,6,6a,6b,7,8,11,11a-decahydro-1H-benzo[a]fluorene-9,2'-3,3a,5,6,7,7a-hexahydrofuro[3,2-b]pyridine]-4'-yl]ethyl]-6-(3-phenylpropanoylamino)hexanamide |
| Industry Type | Forensics |
| Molecular Weight (g/mol) | 700.005 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?