missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy Myristic Acid Methyl Ester, TRC

Product Code. 30390897
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390897 500 mg
30401476 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30390897 Supplier Toronto Research Chemicals Supplier No. TRCH954425500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55682-83-2
Molecular Formula C15 H30 O3
Molecular Weight (g/mol) 258.4
Synonym (±)-Methyl 3-Hydroxytetradecanoate, Methyl 3-Hydroxytetradecanoate, 3-Hydroxytetradecanoic Acid Methyl Ester
IUPAC Name methyl 3-hydroxytetradecanoate
SMILES CCCCCCCCCCCC(O)CC(=O)OC

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C15H30O3/c1-3-4-5-6-7-8-9-10-11-12-14(16)13-15(17)18-2/h14,16H,3-13H2,1-2H3
Formula Weight 258.2195
Chemical Name or Material 3-Hydroxy Myristic Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.