missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-Alpha-methyl-tyrosine Methyl Ester, TRC

Product Code. 30391088
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391088 5 mg -
30398150 50 mg 50mg
30398925 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30391088 Supplier Toronto Research Chemicals Supplier No. TRCH7850105MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 158190-23-9
Molecular Formula C11H15NO4
Molecular Weight (g/mol) 225.24
Synonym 3-Hydroxy-α-Methyl-Tyrosine Methyl Ester
IUPAC Name methyl 2-amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoate
SMILES COC(=O)C(C)(N)Cc1ccc(O)c(O)c1

Specifications

InChI Formula InChI=1S/C11H15NO4/c1-11(12,10(15)16-2)6-7-3-4-8(13)9(14)5-7/h3-5,13-14H,6,12H2,1-2H3
Quantity 5 mg
Formula Weight 225.1001
Shipping Condition Room Temperature
Chemical Name or Material 3-Hydroxy-alpha-methyl-tyrosine Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.