missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-6-chloro-Anagrelide Analogue, TRC

Product Code. 30357524
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357524 10 mg 10mg
30354238 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30357524 Supplier Toronto Research Chemicals Supplier No. TRCD22653010MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Hydroxy-6-chloro-Anagrelide Analogue
Analyte or Component Names 3-Hydroxy-6-chloro-Anagrelide Analogue
Shipping Condition Room Temperature
Molecular Formula C10H8ClN3O2
Synonym 6-Chloro-3-hydroxy-3,5-dihydroimidazo[2,1-b]quinazolin-2(1H)-one
InChI Formula InChI=1S/C10H8ClN3O2/c11-6-2-1-3-7-5(6)4-14-9(16)8(15)13-10(14)12-7/h1-3,9,16H,4H2,(H,12,13,15)
Solution Type Neat
SMILES O=C1NC2=NC3=C(C(Cl)=CC=C3)CN2C1O
IUPAC Name 6-chloro-3-hydroxy-1,5-dihydroimidazo[2,1-b]quinazolin-2(3H)-one
Industry Type Pharmaceutical
Molecular Weight (g/mol) 237.64
Quantity 10 mg
Formula Weight 237.03
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.