missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-5-methylpyridine, 97%

Product Code. 15435407
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15435407 5 g 5g
15445407 250 mg 250mg
15425407 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15435407 Supplier Thermo Scientific Alfa Aesar Supplier No. H32658.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 42732-49-0 | C6H7NO | 109.128 g/mol

3-Hydroxy-5-methylpyridine is used to prepare 3-hydroxy-5-methyl-2-nitropyridine by nitration reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxy-5-methylpyridine is used to prepare 3-hydroxy-5-methyl-2-nitropyridine by nitration reaction.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42732-49-0
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00661297
InChI Key RYJNCIGFPWGVPA-UHFFFAOYSA-N
Synonym 3-hydroxy-5-picoline, 3-hydroxy-5-methylpyridine, 5-methyl-3-hydroxypyridine, 5-hydroxy-3-picoline, 5-methyl-3-pyridinol, 5-methyl-3-pyridol, pubchem6629, 5-methyl-3-hydroxy-pyridine, 3-pyridinol, 5-methyl, 3-hydroxy-5-methylpyridine 250mg
PubChem CID 224753
IUPAC Name 5-methylpyridin-3-ol
SMILES CC1=CC(=CN=C1)O

Specifications

Melting Point 136°C to 140°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 109.13
Percent Purity 97%
Chemical Name or Material 3-Hydroxy-5-methylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.