missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxy-5-methylpyridine, 97%, Thermo Scientific™
£33.10 - £435.00
Chemical Identifiers
CAS | 42732-49-0 |
---|---|
Molecular Formula | C6H7NO |
Molecular Weight (g/mol) | 109.13 |
InChI Key | RYJNCIGFPWGVPA-UHFFFAOYSA-N |
Synonym | 3-hydroxy-5-picoline, 3-hydroxy-5-methylpyridine, 5-methyl-3-hydroxypyridine, 5-hydroxy-3-picoline, 5-methyl-3-pyridinol, 5-methyl-3-pyridol, pubchem6629, 5-methyl-3-hydroxy-pyridine, 3-pyridinol, 5-methyl, 3-hydroxy-5-methylpyridine 250mg |
PubChem CID | 224753 |
IUPAC Name | 5-methylpyridin-3-ol |
SMILES | CC1=CC(=CN=C1)O |
Description
Chemical Identifiers
42732-49-0 | |
109.13 | |
3-hydroxy-5-picoline, 3-hydroxy-5-methylpyridine, 5-methyl-3-hydroxypyridine, 5-hydroxy-3-picoline, 5-methyl-3-pyridinol, 5-methyl-3-pyridol, pubchem6629, 5-methyl-3-hydroxy-pyridine, 3-pyridinol, 5-methyl, 3-hydroxy-5-methylpyridine 250mg | |
5-methylpyridin-3-ol |
C6H7NO | |
RYJNCIGFPWGVPA-UHFFFAOYSA-N | |
224753 | |
CC1=CC(=CN=C1)O |
Specifications
42732-49-0 | |
96.5% min. (GC) | |
C6H7NO | |
Solubility in water: slightly soluble | |
CC1=CC(=CN=C1)O | |
109.13 | |
109.13 | |
3-Hydroxy-5-methylpyridine |
136.0°C to 140.0°C | |
Glass bottle | |
3-hydroxy-5-picoline, 3-hydroxy-5-methylpyridine, 5-methyl-3-hydroxypyridine, 5-hydroxy-3-picoline, 5-methyl-3-pyridinol, 5-methyl-3-pyridol, pubchem6629, 5-methyl-3-hydroxy-pyridine, 3-pyridinol, 5-methyl, 3-hydroxy-5-methylpyridine 250mg | |
RYJNCIGFPWGVPA-UHFFFAOYSA-N | |
5-methylpyridin-3-ol | |
224753 | |
97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 3-Hydroxy-5-methylpyridine
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only