missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-5-methylisoxazole, 97%

Product Code. p-7059348
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11460760

Brand: Thermo Scientific Alfa Aesar 044450.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10004-44-1 | C4H5NO2 | 99.089 g/mol

3-Hydroxy-5-methylisoxazole is used in plant growth stimulation activity and seedlings growing. And it is also used in DNA/RNA synthesis inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10004-44-1
Molecular Formula C4H5NO2
Molecular Weight (g/mol) 99.089
MDL Number MFCD00144468
InChI Key KGVPNLBXJKTABS-UHFFFAOYSA-N
Synonym hymexazol, 3-hydroxy-5-methylisoxazole, hymexazole, 5-methylisoxazol-3-ol, 5-methylisoxazol-3 2h-one, hydroxyisoxazole, tachigaren, itachigarden, 5-methyl-3-hydroxyisoxazole, 5-methyl-3 2h-isoxazolone
PubChem CID 24781
ChEBI CHEBI:5827
IUPAC Name 5-methyl-1,2-oxazol-3-one
SMILES CC1=CC(=O)NO1

Specifications

Quantity 5 g
Merck Index 14,4856
Solubility Information Soluble in alcohol, acetone, THF, chloroform
Formula Weight 99.09
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Hydroxy-5-methylisoxazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.