missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-quinolinecarboxylic acid, 97%

Product Code. 15254794 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15254794 5 g 5g
15244794 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15254794 Supplier Thermo Scientific Acros Supplier No. 449930050
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 118-13-8 | C10H7NO3 | 189.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-13-8
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD01536959
InChI Key MXNVEJDRXSFZQB-UHFFFAOYSA-N
Synonym 3-hydroxy-4-quinolinecarboxylic acid, 3-hydroxycinchoninic acid, cinchoninic acid, 3-hydroxy, 3-hydroxy-quinoline-4-carboxylic acid, 4-quinolinecarboxylic acid, 3-hydroxy, 3-hydroxy-4-quinolinecarboxylicacid, 3-hydroxycinchoninicacid, 4-22-00-02296 beilstein handbook reference, 4-quinolinecarboxylicacid,3-hydroxy, 4-quinolinecarboxylicacid, 3-hydroxy
PubChem CID 8352
IUPAC Name 3-hydroxyquinoline-4-carboxylic acid
SMILES OC(=O)C1=C2C=CC=CC2=NC=C1O

Specifications

Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 189.17
Percent Purity 97%
Chemical Name or Material 3-Hydroxy-4-quinolinecarboxylic acid
compliance-icons
Product Identifier
  • 3-Hydroxy-4-quinolinecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.