missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-Nitroso-2,7- Naphthalenedisulfonic Acid Disodium Salt, Solstice

Product Code. 15666950
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15666950 25 g Glass bottle 25g
15656950 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15666950 Supplier Honeywell-Fluka Supplier No. 7395025G
 Restricted Item
Estimated Shipment: 26-04-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

for spectrophotometric det. of Co, Fe, K, Ag, Ba, Ca, Co, Fe, Ni, Pb

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 525-05-3
Molecular Formula C10H5NNa2O8S2
Molecular Weight (g/mol) 377.25
MDL Number MFCD00149246
InChI Key DMKMTGULLYISBH-UHFFFAOYSA-L
Synonym nitroso-r salt, nitroso r, sodium 3-hydroxy-4-nitrosonaphthalene-2,7-disulfonate, disodium 1-nitroso-2-naphthol-3,6-disulfonate, nitroso r salt, 1-nitroso-3,6-disulfonate, 2,7-naphthalenedisulfonic acid, 3-hydroxy-4-nitroso-, disodium salt, 3-hydroxy-4-nitroso-2,7-naphthalenedisulfonic acid disodium salt, 1-nitroso-2-naphthol-3,6-disulfonic acid disodium salt, disodium 3-hydroxy-4-nitrosonaphthalene-2,7-disulphonate
PubChem CID 10672
IUPAC Name disodium;3-hydroxy-4-nitrosonaphthalene-2,7-disulfonate
SMILES [Na+].[Na+].OC1=C(N=O)C2=CC=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Melting Point 300°C
Color Yellow
Packaging Glass bottle
Quantity 25 g
UN Number NONH for all modes of transport
Beilstein 3643930
Formula Weight 377.26g/mol
Chemical Name or Material 3-Hydroxy-4-nitroso-2,7-naphthalenedisulfonic acid disodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.