missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-nitrobenzoic acid, 98%

Product Code. 11419592
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419592 100 g 100g
11499582 5 g 5g
11409592 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419592 Supplier Thermo Scientific Alfa Aesar Supplier No. A10724.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 619-14-7 | C7H5NO5 | 183.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-14-7
Molecular Formula C7H5NO5
Molecular Weight (g/mol) 183.12
MDL Number MFCD00007110
InChI Key XLDLRRGZWIEEHT-UHFFFAOYSA-N
Synonym 3-hydroxy-4-nitrobenzoic acid, 3-hydroxy-4-nitro-benzoic acid, 4-nitro-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-4-nitro, 3-hydroxy-4-nitrobenzoicacid, pubchem17135, acmc-209mxi, 3-hydroxy4nitrobenzoic acid, ksc356q8j, 3-hydroxy-4-nitro benzoic acid
PubChem CID 69265
IUPAC Name 3-hydroxy-4-nitrobenzoic acid
SMILES OC(=O)C1=CC=C(C(O)=C1)[N+]([O-])=O

Specifications

Melting Point 232°C to 236°C
Quantity 100 g
Beilstein 2107564
Formula Weight 183.12
Percent Purity 98%
Chemical Name or Material 3-Hydroxy-4-nitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.