missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-nitro-3H-isobenzofuran-1-one, Mikromol

Product Code. 30458126
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30458126 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458126 Supplier Mikromol Supplier No. MM3641.190025
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-Hydroxy-4-nitro-3H-isobenzofuran-1-one, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Hydroxy-4-nitro-3H-isobenzofuran-1-one
CAS 39830-64-3
Analyte or Component Names 3-Hydroxy-4-nitro-3H-isobenzofuran-1-one
Shipping Condition Room Temperature
Molecular Formula C8 H5 N O5
Certifications/Compliance ISO 9001
Synonym 1(3H)-Isobenzofuranone, 3-hydroxy-4-nitro- (9CI, ACI), 3-Hydroxy-4-nitro-1(3H)-isobenzofuranone (ACI), 3-Hydroxy-4-nitro-1,3-dihydro-2-benzofuran-1-one, 3-Hydroxy-4-nitro-2-benzofuran-1(3H)-one, 3-Hydroxy-4-nitrophthalide
InChI Formula InChI=1S/C8H5NO5/c10-7-4-2-1-3-5(9(12)13)6(4)8(11)14-7/h1-3,8,11H
Solution Type Neat
SMILES OC1OC(=O)c2cccc(c12)[N+](=O)[O-]
IUPAC Name 3-hydroxy-4-nitro-3H-2-benzofuran-1-one
Molecular Weight (g/mol) 195.129
Quantity 25 mg
Formula Weight 195.0168 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.