missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-methylaniline, TRC

Product Code. 30408225
Change view
Click to view available options
Quantity:
500 mg
5 g
10 g
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30408225 5 g -
30399257 10 g -
30404763 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30408225 Supplier Toronto Research Chemicals Supplier No. TRCH9724155G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2835-95-2
Molecular Formula C7 H9 N O
Molecular Weight (g/mol) 123.15
Synonym o-Cresol, 5-amino- (6CI,7CI,8CI), 5-Amino-2-methylphenol, 1-Methyl-2-hydroxy-4-aminobenzene, 2-Hydroxy-4-aminotoluene, 2-Methyl-5-aminophenol, 3-Amino-6-methylphenol, 3-Hydroxy-4-methylaniline, 3-Hydroxy-p-toluidine, 4-Amino-2-hydroxytoluene, 5-Amino-2-cresol, 5-Amino-o-cresol, 6-Methyl-3-aminophenol
IUPAC Name 5-amino-2-methylphenol
SMILES Cc1ccc(N)cc1O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H9NO/c1-5-2-3-6(8)4-7(5)9/h2-4,9H,8H2,1H3
Formula Weight 123.0684
Chemical Name or Material 5-Amino-2-methylphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.