missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-methoxycinnamic Acid-d3 (Isoferulic Acid-d3), TRC

Product Code. 30391863
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30391863 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30391863 Supplier Toronto Research Chemicals Supplier No. TRCH946182100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material 3-Hydroxy-4-methoxycinnamic Acid-d3 (Isoferulic Acid-d3)
CAS 1028203-97-5
Molecular Formula C10H7D3O4
Quantity 100 mg
Synonym 3-[3-Hydroxy-4-(methoxy-d3)phenyl]acrylic Acid, Isoferulic Acid, 3-[3-Hydroxy-4-(methoxy-d3)phenyl]-2-propenoic Acid
InChI Formula InChI=1S/C10H10O4/c1-14-9-4-2-7(6-8(9)11)3-5-10(12)13/h2-6,11H,1H3,(H,12,13)/b5-3+/i1D3
SMILES OC1=C(OC([2H])([2H])[2H])C=CC(/C=C/C(O)=O)=C1
Molecular Weight (g/mol) 197.2
Formula Weight 197.0767
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.