missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-3-methylbutyric acid, 96%

Product Code. 11365487
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg
Unit Size:
10g
1kg
250g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11365487 250 g 250g
11356149 10 g 10g
11355487 50 g 50g
11375487 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11365487 Supplier Thermo Scientific Alfa Aesar Supplier No. 042722.30
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 625-08-1 | C5H10O3 | 118.13 g/mol

3-Hydroxy-3-methylbutyric acid is a useful biochemical for synthesis and proteomics research. It is used as an indicator of biotin deficiency. Further, it is used to increase muscle mass and decrease muscle breakdown. In addition to this, it is involved in the protein synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-08-1
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00059081
InChI Key AXFYFNCPONWUHW-UHFFFAOYSA-N
Synonym beta-hydroxyisovaleric acid, 3-hydroxyisovaleric acid, 3-hydroxy-3-methylbutyric acid, hmb-d6, 3-oh-isovaleric acid, 3-hydroxy-isovaleric acid, 3-hydroxy-3-methyl-butanoic acid, butanoic acid, 3-hydroxy-3-methyl, b-hydroxyisovaleric acid, beta-hydroxy-beta-methylbutyrate
PubChem CID 69362
ChEBI CHEBI:37084
IUPAC Name 3-hydroxy-3-methylbutanoic acid
SMILES CC(C)(O)CC(O)=O

Specifications

Color Yellow
Density 1.1 g/mL
Boiling Point 128°C (7 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.4408
Quantity 250 g
Beilstein 1743952
Solubility Information Soluble in water and ethanol.
Formula Weight 118.13
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-Hydroxy-3-methylbutyric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.