missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-3-methyl-d3-pentanedioic Acid, TRC

Product Code. 30407665
Change view
Click to view available options
Quantity:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30407665 10 mg
30399460 1 mg
30395966 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30407665 Supplier Toronto Research Chemicals Supplier No. TRCH94826410MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59060-36-5
Molecular Formula C6H7D3O5
Molecular Weight (g/mol) 165.16
Synonym 3-Hydroxy-3-(methyl-d3)pentanedioic Acid
IUPAC Name 3-hydroxy-3-(methyl-d3)pentanedioic acid
SMILES O=C(O)CC(C([2H])([2H])[2H])(O)CC(O)=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C6H10O5/c1-6(11,2-4(7)8)3-5(9)10/h11H,2-3H2,1H3,(H,7,8)(H,9,10)/i1D3
Formula Weight 165.07
Chemical Name or Material Meglutol-D3 (methyl-D3)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.