missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-3-methyl-2-butanone, 90+%

Product Code. 11370328
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11370328 100 g 100g
11390328 5 g 5g
11380328 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11370328 Supplier Thermo Scientific Alfa Aesar Supplier No. 043724.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 115-22-0 | C5H10O2 | 102.133 g/mol

3-Hydroxy-3-methyl-2-butanone used to produce 2,5-dihydroxy-2-methyl-heptan-3-one, laboratory chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-22-0
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00004460
InChI Key BNDRWEVUODOUDW-UHFFFAOYSA-N
Synonym 3-hydroxy-3-methyl-2-butanone, 3-methylacetoin, methylacetoin, 2-butanone, 3-hydroxy-3-methyl, dimethylacetylcarbinol, 2-hydroxy-2-methyl-3-butanone, 2-methyl-2-hydroxybutan-3-one, 1-hydroxy-1-methylethyl methyl ketone, 3-hydroxy-3-methyl-butan-2-one, 3-hydroxy-3-ethylbutanone
PubChem CID 8261
IUPAC Name 3-hydroxy-3-methylbutan-2-one
SMILES CC(=O)C(C)(C)O

Specifications

Density 0.96 g/mL
Boiling Point 140°C to 141°C
Flash Point 42°C (107.6°F)
Refractive Index 1.415
Quantity 100 g
UN Number UN1224
Solubility Information Soluble in chloroform.
Formula Weight 102.13
Percent Purity ≥90%
Physical Form Liquid
Chemical Name or Material 3-Hydroxy-3-methyl-2-butanone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.