missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-3,7,11-trimethyl-1,6,10-dodecatriene, 97+% (total alcohols), Thermo Scientific™

Product Code. 10219133 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10219133 100 g 100g
10408850 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10219133 Supplier Thermo Scientific Acros Supplier No. 121971000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7212-44-4
Molecular Formula C15H26O
Molecular Weight (g/mol) 222.37
MDL Number MFCD00008911,MFCD00008911,MFCD00008911,MFCD00085350
InChI Key FQTLCLSUCSAZDY-SDNWHVSQNA-N
Synonym 3r,6e-nerolidol, unii-uoc0644v25, 3r-6e-nerolidol, nerolidol, 3r,6e-3,7,11-trimethyldodeca-1,6,10-trien-3-ol, e-nerolidol, 1,6,10-dodecatrien-3-ol, 3,7,11-trimethyl-, 3r,6e, nerolidol, 6e--, --nerolidol, ?-nerolidol
PubChem CID 11241545
ChEBI CHEBI:59959
IUPAC Name (3R,6E)-3,7,11-trimethyldodeca-1,6,10-trien-3-ol
SMILES CC(C)=CCC\C(C)=C\CCC(C)(O)C=C

Specifications

Melting Point -75.0°C
Color Yellow
Density 0.8700g/mL
Boiling Point 114.0°C (1.0 mmHg)
Flash Point 96°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Refractive Index 1.4780 to 1.4830
Quantity 100 g
Merck Index 15, 6561
Specific Gravity 0.87
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in abs. alc. and fixed oils, (still clearly sol. in 3 parts of 70% alc.), insoluble in glycerol
Formula Weight 222.37
Percent Purity 97+% (total alcohols)
Physical Form Liquid
Chemical Name or Material 3-Hydroxy-3,7,11-trimethyl-1,6,10-dodecatriene, ≥97% (total alcohols)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Hydroxy-3,7,11-trimethyl-1,6,10-dodecatriene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.