missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-pyrone, TRC

Product Code. 30134635
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134635 50 mg 50mg
30007361 25 mg 25mg
30003249 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134635 Supplier Toronto Research Chemicals Supplier No. TRCH95373350MG
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 496-64-0
Molecular Formula C5 H4 O3
Molecular Weight (g/mol) 112.08
IUPAC Name 3-hydroxypyran-2-one
SMILES OC1=CC=COC1=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C5H4O3/c6-4-2-1-3-8-5(4)7/h1-3,6H
Formula Weight 112.016
Chemical Name or Material 3-Hydroxy-2-pyrone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.