missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-oxobutyl Azilsartan Ester, TRC

Product Code. 30391960
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391960 5 mg 5mg
30395358 25 mg -
30391594 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30391960 Supplier Toronto Research Chemicals Supplier No. TRCH8259605MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1883681-10-4
Molecular Formula C29H26N4O7
Molecular Weight (g/mol) 542.54
Synonym 3-Hydroxy-2-oxobutyl 2-ethoxy-1-((2'-(5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl)-[1,1'-biphenyl]-4-yl)methyl)-1H-benzo[d]imidazole-7-carboxylate
IUPAC Name (3-hydroxy-2-oxobutyl) 2-ethoxy-3-[[4-[2-(5-oxo-2H-1,2,4-oxadiazol-3-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylate
SMILES CCOc1nc2cccc(C(=O)OCC(=O)C(C)O)c2n1Cc3ccc(cc3)c4ccccc4C5=NC(=O)ON5

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C29H26N4O7/c1-3-38-28-30-23-10-6-9-22(27(36)39-16-24(35)17(2)34)25(23)33(28)15-18-11-13-19(14-12-18)20-7-4-5-8-21(20)26-31-29(37)40-32-26/h4-14,17,34H,3,15-16H2,1-2H3,(H,31,32,37)
Formula Weight 542.1801
Chemical Name or Material 3-Hydroxy-2-oxobutyl Azilsartan Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.