missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-nitropyridine, 97%, Thermo Scientific™

Catalog Number 10496970
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
15128-82-2
C5H4N2O3
140.10
MFCD00006258
QBPDSKPWYWIHGA-UHFFFAOYSA-N
3-hydroxy-2-nitropyridine, 2-nitropyridin-3-ol, 2-nitro-3-hydroxypyridine, 2-nitro-3-pyridinol, 3-pyridinol, 2-nitro, 2-nitro-3-hydroxy pyridine, unii-uu357x85yb, 3-hydroxy-2-nitro pyridine, 2-nitro-3-pyridol
27057
2-nitropyridin-3-ol
OC1=CC=CN=C1[N+]([O-])=O
This item is not returnable. View return policy
15128-82-2
C5H4N2O3
140.10
MFCD00006258
QBPDSKPWYWIHGA-UHFFFAOYSA-N
3-hydroxy-2-nitropyridine, 2-nitropyridin-3-ol, 2-nitro-3-hydroxypyridine, 2-nitro-3-pyridinol, 3-pyridinol, 2-nitro, 2-nitro-3-hydroxy pyridine, unii-uu357x85yb, 3-hydroxy-2-nitro pyridine, 2-nitro-3-pyridol
27057
2-nitropyridin-3-ol
OC1=CC=CN=C1[N+]([O-])=O
CAS Min % 96.0
CAS Max % 100.0
Melting Point 67°C to 72°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula HO(C5H3N)NO2
Quantity 100 g
Solubility Information Solubility in water: soluble in water
Formula Weight 140.1
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-2-nitropyridine, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Hydroxy-2-nitropyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'