missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-nitrobenzoic acid, 98+%

Product Code. 11343396
Change view
Click to view available options
Quantity:
500 mg
2 g
10 g
Unit Size:
10g
2g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343396 500 mg 500mg
11353396 2 g 2g
11363396 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11343396 Supplier Thermo Scientific Alfa Aesar Supplier No. L12841.ME

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 602-00-6 | C7H5NO5 | 183.119 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 602-00-6
Molecular Formula C7H5NO5
Molecular Weight (g/mol) 183.119
MDL Number MFCD00017003
InChI Key KPDBKQKRDJPBRM-UHFFFAOYSA-N
Synonym 3-hydroxy-2-nitrobenzoic acid, 3-hydroxy-2-nitrobenzoicacid, 3-hydroxy-2-nitro-benzoic acid, 2-nitro-3-hydroxybenzoic acid, pubchem8846, nitro-m-oxybenzoesaure, benzoic acid, hydroxynitro, ksc288q8j, acmc-1b214, 2-nitro-3-oxidanyl-benzoic acid
PubChem CID 293291
SMILES C1=CC(=C(C(=C1)O)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 177°C to 181°C
Quantity 500 mg
Beilstein 2372248
Formula Weight 183.12
Percent Purity ≥98%
Chemical Name or Material 3-Hydroxy-2-nitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.