missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-naphthoic acid, TRC

Product Code. 30392514
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392514 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392514 Supplier Toronto Research Chemicals Supplier No. TRCH9510781G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Hydroxy-2-naphthoic acid
CAS 92-70-6
Molecular Formula C11 H8 O3
Quantity 1 g
Synonym 2-Naphthoic acid, 3-hydroxy- (8CI), 3-Hydroxy-2-naphthalenecarboxylic acid, 2-Hydroxy-3-carboxynaphthalene, 2-Hydroxy-3-naphthalenecarboxylic acid, 2-Hydroxy-3-naphthoic acid, 2-Hydroxyl-3-naphthoic acid, 2-Naphthol-3-carboxylic acid, 3-Carboxy-2-naphthol, 3-Hydroxy-2-naphthoic acid, 3-Hydroxy-β-naphthoic acid, 3-Naphthol-2-carboxylic acid, BON, BON acid, BONA, C.I. Developer 20, Developer BON, Miketazol Developer ONS, NSC 3719, Naphthol B.O.N., TCI-B 3229, β-Hydroxy-3-naphthoic acid, β-Hydroxynaphthoic acid, β-Oxynaphthoic acid
InChI Formula InChI=1S/C11H8O3/c12-10-6-8-4-2-1-3-7(8)5-9(10)11(13)14/h1-6,12H,(H,13,14)
SMILES OC(=O)c1cc2ccccc2cc1O
IUPAC Name 3-hydroxynaphthalene-2-carboxylic acid
Molecular Weight (g/mol) 188.18
Formula Weight 188.0473
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.