missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-naphthoic acid hydrazide, 98%

Product Code. p-7023112
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453267 25 g 25g
11463267 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11453267 Supplier Thermo Scientific Alfa Aesar Supplier No. A16192.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5341-58-2 | C11H10N2O2 | 202.213 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5341-58-2
Molecular Formula C11H10N2O2
Molecular Weight (g/mol) 202.213
MDL Number MFCD00004097
InChI Key FDNAQCWUERCJBK-UHFFFAOYSA-N
Synonym 3-hydroxy-2-naphthohydrazide, 3-hydroxy-2-naphthoic acid hydrazide, 2-hydroxy-3-naphthoic acid hydrazide, 2-naphthalenecarboxylic acid, 3-hydroxy-, hydrazide, 3-hydroxy-2-naphthoylhydrazine, 2-hydroxy-3-naphthoylhydrazine, 2-naphthoic acid, 3-hydroxy-, hydrazide, 3-hydroxy-2-naphthoic hydrazide, beta-hydroxynaphthoic hydrazide, 3-???naphthalen-2-ol
PubChem CID 72699
IUPAC Name 3-hydroxynaphthalene-2-carbohydrazide
SMILES C1=CC=C2C=C(C(=CC2=C1)C(=O)NN)O

Specifications

Melting Point ∼205°C (decomposition)
Quantity 25 g
Beilstein 395153
Formula Weight 202.21
Percent Purity 98%
Chemical Name or Material 3-Hydroxy-2-naphthoic acid hydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.