missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-methylbutanoic Acid Ethyl Ester(Mixture of Diastereomers), TRC

Product Code. 30392407
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392407 2.5 g 2.5g
30397841 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30392407 Supplier Toronto Research Chemicals Supplier No. TRCH9464902.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27372-03-8
Molecular Formula C7H14O3
Molecular Weight (g/mol) 146.18
Synonym Ethyl 3-Hydroxy-2-methylbutyrate, 3-Hydroxy-2-methyl-butyric Acid Ethyl Ester
IUPAC Name ethyl 3-hydroxy-2-methylbutanoate
SMILES CCOC(=O)C(C)C(C)O

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C7H14O3/c1-4-10-7(9)5(2)6(3)8/h5-6,8H,4H2,1-3H3
Formula Weight 146.0943
Chemical Name or Material 3-Hydroxy-2-methylbutanoic Acid Ethyl Ester(Mixture of Diastereomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.