missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-methyl-d3-benzoic Acid, TRC

Product Code. 30392801
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30392801 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30392801 Supplier Toronto Research Chemicals Supplier No. TRCH946407100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Hydroxy-2-methyl-d3-benzoic Acid
CAS 1020719-51-0
Molecular Formula C8 2H3 H5 O3
Quantity 100 mg
Synonym Benzoic acid, 3-hydroxy-2-(methyl-d3)-, 3,2-Cresotic acid-d3, 3-Hydroxy-2-(methyl-d3)benzoic acid, 3-Hydroxy-2-methyl-d3-benzoic acid, 2-(Methyl-d3)-3-hydroxybenzoic acid, 3-Hydroxy-2-methylbenzoic acid D3 (methyl-D3)
InChI Formula InChI=1S/C8H8O3/c1-5-6(8(10)11)3-2-4-7(5)9/h2-4,9H,1H3,(H,10,11)/i1D3
SMILES [2H]C([2H])([2H])c1c(O)cccc1C(=O)O
IUPAC Name 3-hydroxy-2-(trideuteriomethyl)benzoic acid
Molecular Weight (g/mol) 155.17
Formula Weight 155.0662
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.