missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-methyl-4-pyrone, 99%

Product Code. 10205070 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10205070 100 g Glass bottle 100g
10456812 5 g Glass bottle 5g
10520951 25 g Glass bottle 25g
10494925 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10205070 Supplier Thermo Scientific Acros Supplier No. 121551000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 118-71-8 | C6H6O3 | 126.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-71-8
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00006578
InChI Key XPCTZQVDEJYUGT-UHFFFAOYSA-N
Synonym maltol, 3-hydroxy-2-methyl-4-pyrone, 3-hydroxy-2-methyl-4h-pyran-4-one, larixinic acid, talmon, palatone, vetol, larixic acid, corps praline, 4h-pyran-4-one, 3-hydroxy-2-methyl
PubChem CID 8369
ChEBI CHEBI:69438
SMILES CC1=C(O)C(=O)C=CO1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 160°C to 164°C
Color Colorless
Boiling Point 170°C (100.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5 % min. (GC)
Packaging Glass bottle
Quantity 100 g
Merck Index 15, 5777
Solubility Information Solubility in water: 12g/L (25°C). Other solubilities: soluble in alcohol and propylene glycol,sparingly soluble in benzene and ether
IUPAC Name 3-hydroxy-2-methylpyran-4-one
Formula Weight 126.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Hydroxy-2-methyl-4-pyrone
Show More Show Less
compliance-icons
Product Identifier
  • 3-Hydroxy-2-methyl-4-pyrone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.