missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2'-methoxy-2-naphthanilide, TRC

Product Code. 30394317
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394317 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30394317 Supplier Toronto Research Chemicals Supplier No. TRCH9730802.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Hydroxy-2'-methoxy-2-naphthanilide
CAS 135-62-6
Molecular Formula C18H15NO3
Quantity 2.5 g
InChI Formula InChI=1S/C18H15NO3/c1-22-17-9-5-4-8-15(17)19-18(21)14-10-12-6-2-3-7-13(12)11-16(14)20/h2-11,20H,1H3,(H,19,21)
SMILES COc1ccccc1NC(=O)c2cc3ccccc3cc2O
IUPAC Name 3-hydroxy-N-(2-methoxyphenyl)naphthalene-2-carboxamide
Molecular Weight (g/mol) 293.31
Formula Weight 293.1052
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.