missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-iodopyridine, 97%, Thermo Scientific™

Product Code. 10881881 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10881881 1 g Glass bottle 1g
10697220 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10881881 Supplier Thermo Scientific Acros Supplier No. 436390010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 40263-57-8
Molecular Formula C5H4INO
Molecular Weight (g/mol) 220.99
InChI Key HJBGMPCMSWJZNH-UHFFFAOYSA-N
Synonym 2-iodo-3-hydroxypyridine, 2-iodo-3-pyridinol, 3-hydroxy-2-iodopyridine, 3-pyridinol, 2-iodo, 2-iodo-pyridin-3-ol, 2-iodpyridin-3-ol, 2-iodopyridine-3-ol, acmc-209jcf, 2-iodo-3-hydroxypyridin, 2-iodo-3-hydroxy-pyridine
PubChem CID 97179
IUPAC Name 2-iodopyridin-3-ol
SMILES C1=CC(=C(N=C1)I)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 191.4°C to 193.2°C
Color Tan to White
Elemental Analysis 26.08 to 28.25% C, 6.09 to 6.59% N
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 220.99
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-2-iodopyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Iodo-3-hydroxypyridine, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.