missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2,4,6-triiodobenzoic acid, 97%

Product Code. 11440967
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440967 100 g 100g
11420967 5 g 5g
11430967 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440967 Supplier Thermo Scientific Alfa Aesar Supplier No. B23355.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 53279-72-4 | C7H3I3O3 | 515.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53279-72-4
Molecular Formula C7H3I3O3
Molecular Weight (g/mol) 515.81
MDL Number MFCD00020305
InChI Key GIAVHGFPMPSIFI-UHFFFAOYSA-N
Synonym htba, 2,4,6-triiodo-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-2,4,6-triiodo, 3-hydroxy-2,4,6-triiodobenzoicacid, pubchem17410, acmc-1aozs, ksc496i6r, 3 hydroxy-2,4,6-triiodobenzoic acid, 3-hydroxy-2,4,6 triiodobenzoic acid
PubChem CID 96627
IUPAC Name 3-hydroxy-2,4,6-triiodobenzoic acid
SMILES OC(=O)C1=C(I)C=C(I)C(O)=C1I

Specifications

Melting Point 211°C to 215°C
Quantity 100 g
Beilstein 2113494
Sensitivity Light sensitive
Formula Weight 515.81
Percent Purity 97%
Chemical Name or Material 3-Hydroxy-2,4,6-triiodobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.