missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2,2-dimethylpropionic acid, 97+%

Product Code. 11373406
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373406 50 g 50g
11383406 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11373406 Supplier Thermo Scientific Alfa Aesar Supplier No. L12858.18
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 4835-90-9 | C5H10O3 | 118.132 g/mol

3-Hydroxy-2,2-dimethylpropionic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxy-2,2-dimethylpropionic acid is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4835-90-9
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00059953
InChI Key RDFQSFOGKVZWKF-UHFFFAOYSA-N
Synonym hydroxypivalic acid, 3-hydroxypivalic acid, 2,2-dimethyl-3-hydroxypropionic acid, propanoic acid, 3-hydroxy-2,2-dimethyl, 3-hydroxy-2,2-dimethylpropionic acid, unii-bjp2cxk56p, bjp2cxk56p, acmc-1arbc, beta-hydroxy pivalic acid
PubChem CID 78548
IUPAC Name 3-hydroxy-2,2-dimethylpropanoic acid
SMILES CC(C)(CO)C(=O)O

Specifications

Melting Point 122°C to 126°C
Odor Odorless
Quantity 50 g
Beilstein 1747025
Solubility Information Soluble in water.
Formula Weight 118.13
Percent Purity 98%
Chemical Name or Material 3-Hydroxy-2,2-dimethylpropionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.