missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-1-methylpiperidine, 98%, Thermo Scientific Chemicals

Codice prodotto. 11393006
Cambia vista
Click to view available options
Quantity:
5 g
25 g
Dimensione della confezione:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Dimensione della confezione
11393006 5 g 5g
11303016 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Normativo: Possono essere applicate restrizioni all'acquisto di questo prodotto e potremmo avere bisogno di richiedere ulteriore documentazione all'utente.
Codice prodotto. 11393006 Fornitore Thermo Scientific Alfa Aesar N. del fornitore L10829.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Normativo: Possono essere applicate restrizioni all'acquisto di questo prodotto e potremmo avere bisogno di richiedere ulteriore documentazione all'utente.

Reactant for Optimization of Novobiocin scaffold to produce antitumor agents, Formation of carbamates and N-alkylimidazoles. Reactant for synthesis of Pyridine derivatives as CDK5 inhibitors Phenylcarbamate derivatives as ligands for nicotinic acetylcholine receptors, Amino phosphite ligands, Mexiletine enantiomers by nucleophilic aromatic substitution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant for Optimization of Novobiocin scaffold to produce antitumor agents, Formation of carbamates and N-alkylimidazoles. Reactant for synthesis of Pyridine derivatives as CDK5 inhibitors Phenylcarbamate derivatives as ligands for nicotinic acetylcholine receptors, Amino phosphite ligands, Mexiletine enantiomers by nucleophilic aromatic substitution.

Solubility
Fully misicible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3554-74-3
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00006495
InChI Key UKANCZCEGQDKGF-UHFFFAOYSA-N
Synonym 3-hydroxy-1-methylpiperidine, 1-methyl-3-piperidinol, n-methyl-3-piperidinol, 3-piperidinol, 1-methyl, 3-hydroxy-n-methylpiperidine, 1-methyl-3-hydroxypiperidine, n-methyl-3-hydroxypiperidine, n-methyl-3-hydroxy piperidine, 3r-1-methyl-3-piperidinol, pubchem6759
PubChem CID 98016
IUPAC Name 1-methylpiperidin-3-ol
SMILES CN1CCCC(C1)O

Specifications

Density 0.999
Boiling Point 76°C to 78°C (11 mmHg)
Flash Point 70°C (158°F)
Refractive Index 1.474
Quantity 5 g
Beilstein 103017
Solubility Information Fully misicible in water.
Formula Weight 115.18
Percent Purity 98%
Chemical Name or Material 3-Hydroxy-1-methylpiperidine
compliance-icons
Product Identifier
  • 3-Hydroxy-1-methylpiperidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 3-Hydroxy-1-methylpiperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.