missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-1-methylpiperidine, 97%, Thermo Scientific Chemicals

Product Code. 15244614 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15244614 5 g 5g
15254614 25 g 25g
15274614 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15244614 Supplier Thermo Scientific Acros Supplier No. 121500050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3554-74-3
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.17
InChI Key UKANCZCEGQDKGF-UHFFFAOYSA-N
Synonym 3-hydroxy-1-methylpiperidine, 1-methyl-3-piperidinol, n-methyl-3-piperidinol, 3-piperidinol, 1-methyl, 3-hydroxy-n-methylpiperidine, 1-methyl-3-hydroxypiperidine, n-methyl-3-hydroxypiperidine, n-methyl-3-hydroxy piperidine, 3r-1-methyl-3-piperidinol, pubchem6759
PubChem CID 98016
IUPAC Name 1-methylpiperidin-3-ol
SMILES CN1CCCC(C1)O

Specifications

Density 0.9990g/mL
Boiling Point 76.0°C to 78.0°C (11.0 mmHg)
Flash Point 70°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.474
Quantity 5 g
Specific Gravity 0.999
Formula Weight 115.17
Percent Purity 97%
Chemical Name or Material 3-Hydroxy-1-methylpiperidine
Show More Show Less
compliance-icons
Product Identifier
  • 3-Hydroxy-N-methylpiperidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.