missing translation for 'onlineSavingsMsg'
Learn More

3-Hexylthiophene, 98%

Product Code. 10190032 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10190032 5 g Glass bottle 5g
10195633 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10190032 Supplier Thermo Scientific Acros Supplier No. 367380050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1693-86-3 | C10H16S | 168.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1693-86-3
Molecular Formula C10H16S
Molecular Weight (g/mol) 168.3
MDL Number MFCD00143181
InChI Key JEDHEMYZURJGRQ-UHFFFAOYSA-N
Synonym 3-n-hexylthiophene, thiophene, 3-hexyl, 3-hexylthiopene, 3-hexylthiophen, 4-hexylthiophene, hexylthiophene 3-, pubchem8861, poly, 3-hexylthiophene, 3-hexylthiophene, 97+%
PubChem CID 566849
IUPAC Name 3-hexylthiophene
SMILES CCCCCCC1=CSC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow or Red
Density 0.9360g/mL
Boiling Point 65.0°C (0.4 mmHg)
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4945 to 1.4975
Quantity 5 g
Specific Gravity 0.936
Formula Weight 168.3
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Hexylthiophene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.