missing translation for 'onlineSavingsMsg'
Learn More

3-Hexylthiophene, 98%, Thermo Scientific Chemicals

Catalog Number p-2690393
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1693-86-3
C10H16S
168.3
MFCD00143181
JEDHEMYZURJGRQ-UHFFFAOYSA-N
3-n-hexylthiophene, thiophene, 3-hexyl, 3-hexylthiopene, 3-hexylthiophen, 4-hexylthiophene, hexylthiophene 3-, pubchem8861, poly, 3-hexylthiophene, 3-hexylthiophene, 97+%
566849
3-hexylthiophene
CCCCCCC1=CSC=C1
This item is not returnable. View return policy
1693-86-3
C10H16S
168.3
MFCD00143181
JEDHEMYZURJGRQ-UHFFFAOYSA-N
3-n-hexylthiophene, thiophene, 3-hexyl, 3-hexylthiopene, 3-hexylthiophen, 4-hexylthiophene, hexylthiophene 3-, pubchem8861, poly, 3-hexylthiophene, 3-hexylthiophene, 97+%
566849
3-hexylthiophene
CCCCCCC1=CSC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow or Red
Density 0.9360g/mL
Boiling Point 65.0°C (0.4 mmHg)
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4945 to 1.4975
Quantity 5 g
Specific Gravity 0.936
Formula Weight 168.3
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Hexylthiophene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'