missing translation for 'onlineSavingsMsg'
Learn More

3-Hexanol, 98%

Product Code. 11499656
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499656 25 g 25g
11409666 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499656 Supplier Thermo Scientific Alfa Aesar Supplier No. B20210.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 623-37-0 | C6H14O | 102.177 g/mol

It finds it application as a flavoring agent or adjuvant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it application as a flavoring agent or adjuvant.

Solubility
Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-37-0
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00004582
InChI Key ZOCHHNOQQHDWHG-UHFFFAOYSA-N
Synonym 3-hexanol, ethyl propyl carbinol, 3-hexyl alcohol, 3-hexanol natural, ethylpropylcarbinol, fema no. 3351, hexanol-3, 1-butanol, ethyl, acmc-209n2d
PubChem CID 12178
IUPAC Name hexan-3-ol
SMILES CCCC(CC)O

Specifications

Density 0.81
Boiling Point 134°C to 136°C
Flash Point 42°C (108°F)
Odor Characteristic
Refractive Index 1.415
Quantity 25 g
UN Number UN2282
Beilstein 1718964
Solubility Information Soluble in alcohol.
Formula Weight 102.18
Percent Purity 98%
Chemical Name or Material 3-Hexanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Hexanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 3-Hexanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.